Learn More
Thermco Products Inc Single Probe - Pt100 High Precision Digital Thermometer
Catalog No. NC1685642
Encompass_Preferred
Handheld Pt100 Thermometers are High Precision, Metrology Grade, NIST Traceable Instruments. These high accuracy thermometers are used when temperature measurement are critical and/or when online documentation is required.
Catalog No. NC1685642 Supplier Thermco Products Inc Supplier No. ACCD750P
Handheld Pt100 Thermometers are High Precision, Metrology Grade, NIST Traceable Instruments. These high accuracy thermometers are used when temperature measurement are critical and/or when online documentation is required.