Learn More
Thermco Products Inc PT100 IMM PROBE 150X3MM -200
Catalog No. NC1252629
Encompass_Preferred
High Precision Pt100 Probes, �0.03�C (-30 to +200�C) - 1 Meter Cable PVC
Catalog No. NC1252629 Supplier Thermco Products Inc Supplier No. ACCD1018
High Precision Pt100 Probes, �0.03�C (-30 to +200�C) - 1 Meter Cable PVC