Learn More
Thermco Products Inc Handheld Pt100 Digital Thermometer Nist Traceable Laboratory Temperature Standard
Catalog No. NC0737040
Encompass_Preferred
PRECISION Handheld Pt100 Digital Thermometer for the verification and calibration of liquid-in-glass, digital and dial bi-metal thermometer; as well as laboratory instruments such as freezers, refrigerators, incubators, ovens, etc. The PRECISION - Pt100’s are traceable to NIST Standards and is furnished with a certificate proving traceability. 100/300 C&F x 0.1 C&F, Certified @ -20, 0, 4, 25, 37, 56, 95, 121C
Catalog No. NC0737040 Supplier Thermco Products Inc Supplier No. ACCD370PFC
PRECISION Handheld Pt100 Digital Thermometer for the verification and calibration of liquid-in-glass, digital and dial bi-metal thermometer; as well as laboratory instruments such as freezers, refrigerators, incubators, ovens, etc. The PRECISION - Pt100’s are traceable to NIST Standards and is furnished with a certificate proving traceability. 100/300 C&F x 0.1 C&F, Certified @ -20, 0, 4, 25, 37, 56, 95, 121C