Learn More
Elpro PT100 PROBE,TEMP -50 TO 105C
SDP
Catalog No. NC2242692
Encompass
Pt100 probe, temperature range -50 °C..+105 °C, cable length 2.65 m, plastic material, waterproof, with M8 connector to connect to LIBERO CE/GEPRO_PT100_P20D5_PLA2.65_M8
This item is not returnable. View return policy
Catalog No. NC2242692 Supplier Elpro Supplier No. 802291
Pt100 probe, temperature range -50 °C..+105 °C, cable length 2.65 m, plastic material, waterproof, with M8 connector to connect to LIBERO CE/GEPRO_PT100_P20D5_PLA2.65_M8