Learn More
Chemglass Life Sciences PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/4" OD x 18" Long, Style "B", Fits Reactor Capacity: 1-5L
SDP
Catalog No. NC1288863
Encompass_Preferred
Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/4" OD x 18" Long, Style "B", Fits Reactor Capacity: 1-5L
Catalog No. NC1288863 Supplier Chemglass Life Sciences Supplier No. CG1978P918
Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/4" OD x 18" Long, Style "B", Fits Reactor Capacity: 1-5L