Learn More
Chemglass Life Sciences PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/4" OD x 12" Long, Style "B", Fits Reactor Capacity: 500mL
SDP
Catalog No. NC1311876
Encompass_Preferred
pt100 sensor, pfa coated, 1/4"od x 12" long style b, fits reactor capacity 500ml
Catalog No. NC1311876 Supplier Chemglass Life Sciences Supplier No. CG1978P912
pt100 sensor, pfa coated, 1/4"od x 12" long style b, fits reactor capacity 500ml