Learn More
Julabo USA Inc Pt100 sensor, 24" x 1/4", stainless steel, 12' cable with Lemosa connector
SDP
Catalog No. 502068324
Encompass_Preferred
Pt100 temperature sensor, stainless steel; 24" long x 1/4" OD with 12' cable and 4-pin Lemosa connector. For use with any JULABO circulator having external Pt100 sensor capability. Shielded cable.
Catalog No. 50-206-8324 Supplier Julabo USA Inc Supplier No. 8891001
Pt100 temperature sensor, stainless steel; 24" long x 1/4" OD with 12' cable and 4-pin Lemosa connector. For use with any JULABO circulator having external Pt100 sensor capability. Shielded cable.