Learn More
Chemglass Life Sciences PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/4" OD x 32" Long, Style "B", Fits Reactor Capacity: 30L
SDP
Catalog No. NC1282130
Encompass_Preferred
PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/4" OD x 32" Long, Style "B", Fits Reactor Capacity: 30L
Catalog No. NC1282130 Supplier Chemglass Life Sciences Supplier No. CG1978P932
PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/4" OD x 32" Long, Style "B", Fits Reactor Capacity: 30L