Learn More
Chemglass Life Sciences PT100 Sensor, 1/2" OD x 45" Lgth FEP Clear Coated, 12ft lead, Green Shielded Cable, Lemo Connector, Strain Relief Spring, Style "B", Fits Reactor Capacity: 50L (Tall)
SDP
Catalog No. 501531364
Encompass_Preferred
PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/2" OD x 45" Long, Style "B", Fits Reactor Capacity: 50L (Tall). PT100 RTD temperature sensor is for direct insertion into flasks and process reactors using any O-Ring style compression adapter, such as the CG-1042-E. Probes are constructed using shielded cable to ensure a clean signal. Style B has a Lemo Connector direct wired to a 12 foot lead and is for use with all Huber circulators permitting process temperature control based on the contents of the reactor. Sensor is constructed using GREEN shielded wire/cable. Lemo connector can be used as a ground for the probe using an optional grounding clip and wire. An adapter having a female Lemo connector to the 2-blade mini plug is available to allow use of the Style B sensors with equipment having this connection.
Catalog No. 50-153-1364 Supplier Chemglass Life Sciences Supplier No. CG1978P946
PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/2" OD x 45" Long, Style "B", Fits Reactor Capacity: 50L (Tall). PT100 RTD temperature sensor is for direct insertion into flasks and process reactors using any O-Ring style compression adapter, such as the CG-1042-E. Probes are constructed using shielded cable to ensure a clean signal. Style B has a Lemo Connector direct wired to a 12 foot lead and is for use with all Huber circulators permitting process temperature control based on the contents of the reactor. Sensor is constructed using GREEN shielded wire/cable. Lemo connector can be used as a ground for the probe using an optional grounding clip and wire. An adapter having a female Lemo connector to the 2-blade mini plug is available to allow use of the Style B sensors with equipment having this connection.