Learn More
Chemglass Life Sciences Pt100 Temperature Probe, AISI 316 Ti Ø3mm
SDP
Catalog No. 5000493072
Encompass_Preferred
Pt100 Temperature Probe AISI 316 Ti diameter 3mm. For use with CG-1991-V ECODryBlock Systems / Dryblock Heaters.
Catalog No. 50-004-93072 Supplier Chemglass Life Sciences Supplier No. CG1991V200
Pt100 Temperature Probe AISI 316 Ti diameter 3mm. For use with CG-1991-V ECODryBlock Systems / Dryblock Heaters.