Learn More
Chemglass Life Sciences PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/8" OD x 12" Long, Style "A"
SDP
Catalog No. 501531360
Encompass_Preferred
PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/8" OD x 12" Long, Style "A". PT100 RTD temperature sensor is for direct insertion into flasks and process reactors using any O-Ring style compression adapter, such as the CG-1042-E. Probes are constructed using shielded cable to ensure a clean signal. Style A has a male lemo connector and can be used with CG-3499-10 white coiled extension cords which have a female lemo connection on one end and a male mini plug on the other. Probe is constructed using shielded wire/cable. These will fit the J-Kem temperature controllers (CG-3200 through CG-3210). Probes are designed to be used with round bottom flasks.
Catalog No. 50-153-1360 Supplier Chemglass Life Sciences Supplier No. CG1978P902
PT100 Sensor, Constructed with Shielded Cable, the Lemo Connector Provides An Area for a Ground Connection, PFA Coated, 1/8" OD x 12" Long, Style "A". PT100 RTD temperature sensor is for direct insertion into flasks and process reactors using any O-Ring style compression adapter, such as the CG-1042-E. Probes are constructed using shielded cable to ensure a clean signal. Style A has a male lemo connector and can be used with CG-3499-10 white coiled extension cords which have a female lemo connection on one end and a male mini plug on the other. Probe is constructed using shielded wire/cable. These will fit the J-Kem temperature controllers (CG-3200 through CG-3210). Probes are designed to be used with round bottom flasks.