Learn More
Selleck Chemical LLC PT2385 1mg 1672665-49-4
SDP
Supplier:  Selleck Chemical LLC S83521MG
Encompass_Preferred
PT2385 is a HIF-2? antagonist with luciferase EC50 of 27 nM and no significant off-target activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8293