Learn More
Medchemexpress LLC HY-125840 5mg Medchemexpress, PT2977 CAS:1672668-24-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1258405MG
Encompass_Preferred
Medchemexpress, HY-125840 5mg PT2977 CAS:1672668-24-4 PT2977 (MK-6482) is an orally active and selective HIF-2α inhibitor with an IC50 of 9 nM. PT2977, as a second-generation HIF-2α inhibitor, increases potency and improves pharmacokinetic profile. PT2977 is a potential treatment for clear cell renal cell carcinoma (ccRCC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8894