Learn More
IHEALTH LABS INC iHealth Wireless Non-Contact Infrared Forehead Thermometer
SDP
Catalog No. NC3451876
Encompass_Preferred
Take control of your health with the iHealth Wireless No-Touch Forehead Thermometer PT3SBT.lt's important to know each individual's normal temperature when they are well. This is the only way to accurately diagnose a fever.1. Measure and share temperature results, and track yourimprovement over time.2. Compatible with iOS and AndroidSpecifications-model- PT3SBTPower supply - 2 X 1.5V AAA size alkaline batteriesDisplay resolution - 0.1°F or CWeight- 70g (exclude batteries)
This item is not returnable. View return policy
Catalog No. NC3451876 Supplier IHEALTH LABS INC Supplier No. PT3SBT
Take control of your health with the iHealth Wireless No-Touch Forehead Thermometer PT3SBT.lt's important to know each individual's normal temperature when they are well. This is the only way to accurately diagnose a fever.1. Measure and share temperature results, and track yourimprovement over time.2. Compatible with iOS and AndroidSpecifications-model- PT3SBTPower supply - 2 X 1.5V AAA size alkaline batteriesDisplay resolution - 0.1°F or CWeight- 70g (exclude batteries)