Learn More
AdipoGen PTACH
SDP
Catalog No. 502560262
Encompass_Preferred
Chemical. CAS 848354-66-5. Formula C20H26N2O2S2. MW 390.56. Synthetic. Non-hydroxamate HDAC inhibitor HDACi IC50 32, 48 and 41nM for HDAC4, HDAC1 and HDAC6, respectively. Cell-permeable prodrug that is intracellularly converted to a potent HDAC inhibitor. Predicted to exhibit a similar HDAC binding mode as that of SAHA, interacting with the active-site zinc targeting group. Shown to exhibit comparable antiproliferative and apoptotic activity as SAHA against various cancer cell lines. Inhibits growth of various cancer cells in vitro EC50 = 1.1 - 9.1mM. Also reactivates latent HIV-1 gene expression.
Catalog No. 50-256-0262 Supplier AdipoGen Supplier No. CDXP0063M005
Chemical. CAS 848354-66-5. Formula C20H26N2O2S2. MW 390.56. Synthetic. Non-hydroxamate HDAC inhibitor HDACi IC50 32, 48 and 41nM for HDAC4, HDAC1 and HDAC6, respectively. Cell-permeable prodrug that is intracellularly converted to a potent HDAC inhibitor. Predicted to exhibit a similar HDAC binding mode as that of SAHA, interacting with the active-site zinc targeting group. Shown to exhibit comparable antiproliferative and apoptotic activity as SAHA against various cancer cell lines. Inhibits growth of various cancer cells in vitro EC50 = 1.1 - 9.1mM. Also reactivates latent HIV-1 gene expression.