Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PTAD-PEG8-alkyne | 684.73 | 5 MG
SDP

Catalog No. 5000391443
Encompass_Preferred

PTAD-PEG8-alkyne is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).

  • Used as a cleavable ADC linker
  • Target: ADC linker
  • Pathway: Antibody-drug conjugate/ADC related
  • Storage: Pure form: -20°C for 3 years
  • Storage: In solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-91443 Supplier Medchemexpress LLC Supplier No. HY1649285MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PTAD-PEG8-alkyne is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).

  • Used as a cleavable ADC linker
  • Target: ADC linker
  • Pathway: Antibody-drug conjugate/ADC related
  • Storage: Pure form: -20°C for 3 years
  • Storage: In solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.