Learn More
Medchemexpress LLC PTAD-PEG8-azide | 99.77% | 715.75 | 5 MG
SDP
Catalog No. 5000391750
Encompass_Preferred
PTAD-PEG8-azide is a cleavable ADC linker containing an azide group. It can be used in the synthesis of antibody-drug conjugates (ADCs).
  • Cleavable ADC linker
  • Contains an azide group
  • Used in the synthesis of antibody-drug conjugates (ADCs)
Catalog No. 50-003-91750 Supplier Medchemexpress LLC Supplier No. HY1649295MG
PTAD-PEG8-azide is a cleavable ADC linker containing an azide group. It can be used in the synthesis of antibody-drug conjugates (ADCs).
  • Cleavable ADC linker
  • Contains an azide group
  • Used in the synthesis of antibody-drug conjugates (ADCs)