Learn More
Diagnostic Biosystems PTAH PTAH
Catalog No. 502407313
Encompass_Preferred
The PTAH Stain Kit is intended for use in the histological visualization of collagen, striated muscle, glial fibers and collagen without using Zenker’s Fixative containing Mercuric Chloride. This kit may be used on formalin-fixed, paraffin-embedded or frozen sections.
Catalog No. 50-240-7313 Supplier Diagnostic Biosystems Supplier No. KT029
The PTAH Stain Kit is intended for use in the histological visualization of collagen, striated muscle, glial fibers and collagen without using Zenker’s Fixative containing Mercuric Chloride. This kit may be used on formalin-fixed, paraffin-embedded or frozen sections.