Learn More
Newcomer Supply Phosphotungstic Acid Hematoxylin (PTAH) Stain Kit, 250 ml kit
SDP
Catalog No. NC2102261
Encompass
Newcomer Supply Phosphotungstic Acid Hematoxylin (PTAH) Stain Kit procedure, with included microwave modification, is used for the demonstration of collagen, muscle striations and central nervous system (CNS) structures. The Zenker fixative used in this procedure is modified with zinc chloride and does not contain mercury.
Catalog No. NC2102261 Supplier Newcomer Supply Supplier No. 9111A
Newcomer Supply Phosphotungstic Acid Hematoxylin (PTAH) Stain Kit procedure, with included microwave modification, is used for the demonstration of collagen, muscle striations and central nervous system (CNS) structures. The Zenker fixative used in this procedure is modified with zinc chloride and does not contain mercury.