Learn More
Adam Equipment PTB 2602e - Polaris Precision Balances - Capacity: 2600g, Readability: 0.01g
Supplier:  Adam Equipment PTB 2602E
Encompass_Preferred
Polaris Precision Balances deliver accuracy and efficiency for laboratory, research, and industrial applications. A large display with fast stabilization enhances productivity, while the color-changing backlight provides clear checkweighing feedback to ensure consistent, accurate results.Advanced connectivity—including Bluetooth, RJ45 Ethernet, RS-232, and USB—enables seamless integration with peripherals, improving data management. GLP-compliant output with date and time stamps ensures reliable recordkeeping for regulated environments.A grade 304 stainless steel pan allows easy cleaning and maintains hygiene, while a Kensington-type lock slot adds security in shared spaces.With intuitive operation, robust connectivity, and dependable performance, Polaris Precision Balances are a reliable solution for precise, efficient weighing.
Catalog No. NC3965067