Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ptb7

Catalog No. 772410100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-241-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-241-0100MG Supplier Sigma Aldrich Supplier No. 772410100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PTB7 is a semiconducting polymer used in organic photovoltaics with an energy efficiency of 9.15%. It can act as an electron donor with narrow optical band gaps and excellent conjugation while forming a nanocomposite with fullerenes.It is majorly used as an active layer that enhances the overall performance by increasing the light absorption and improving the electron mobility of polymeric solar cells (PSCs).

Specifications

CAS 1266549-31-8
Molecular Formula (C41H53FO4S4)n
Linear Formula (C41H53FO4S4)n
Quantity 100 mg
Synonym Poly({4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b]dithiophene-2,6-diyl}{3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl})
Molecular Weight (g/mol) 80,000-200,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.