Learn More
Aviva Systems Biology Corp PTBP2 Antibody - N-terminal region (ARP40966_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40966_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PTBP2. Alternate names: FLJ34897, MIBP, PTB, PTBLP, brPTB, nPTB, nPTB5, nPTB6, nPTB7, nPTB8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57679