Learn More
Strem, An Ascensus Company CAS# 166445-62-1. 1g. Di-mu-iodobis(tri-t-butylphosphino)dipalladium(I), 98%.
SDP
Supplier:  Strem, An Ascensus Company 4603101G
Encompass
CAS# 166445-62-1. 1g. Di-mu-iodobis(tri-t-butylphosphino)dipalladium(I), 98%. . Molecular Weight: 871.28. Molecular Formula: C24H54I2P2Pd2. Color/form: black solid Strem# 46-0310. http://www.strem.com/catalog/v/46-0310/
Catalog No. 50-201-6776