Learn More
Apexbio Technology LLC PTC-209 315704-66-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062729
Encompass_Preferred
PTC-209 (CAS 315704-66-6) is a small-molecule inhibitor targeting BMI-1 identified through Gene Expression Modulation by Small molecules (GEMS) technology It acts by suppressing BMI-1 expression at the transcriptional level displaying an IC50 of approximately 0 5 M In HCT116 cells PTC-209 reduces both UTR-mediated and endogenous BMI-1 expression It selectively restricts cell proliferation in certain tumor and stem cell types including U937 HT1080 and human hematopoietic stem cells without affecting viability in cell lines such as HEK293 and HT1080 Thus PTC-209 is primarily utilized in studies exploring cancer stem cell growth and oncogenesis involving BMI-1 and the PRC1 complex
Catalog No. 50-000-62729 Supplier Apexbio Technology LLC Supplier No. B31795.1
PTC-209 (CAS 315704-66-6) is a small-molecule inhibitor targeting BMI-1 identified through Gene Expression Modulation by Small molecules (GEMS) technology It acts by suppressing BMI-1 expression at the transcriptional level displaying an IC50 of approximately 0 5 M In HCT116 cells PTC-209 reduces both UTR-mediated and endogenous BMI-1 expression It selectively restricts cell proliferation in certain tumor and stem cell types including U937 HT1080 and human hematopoietic stem cells without affecting viability in cell lines such as HEK293 and HT1080 Thus PTC-209 is primarily utilized in studies exploring cancer stem cell growth and oncogenesis involving BMI-1 and the PRC1 complex