Learn More
Medchemexpress LLC HY-15888 5mg , PTC-209 CAS:315704-66-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY158885MG
Encompass_Preferred
HY-15888 5mg PTC-209 CAS: 315704-66-6 PTC-209 is a specific BMI-1 inhibitor with an IC50 of 0.5 μM in HEK293T cell line. PTC-209 irreversibly impairs colorectal cancer-initiating cells (CICs). PTC-209 shows potent anti-myeloma activity and impairs the tumor microenvironment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1881