Learn More
Medchemexpress LLC PTC-209 hydrobromide | 1217022-63-3 | 99.0% | 576.10 | C17H14Br3N5OS | 10 MG
SDP
Catalog No. 5000288312
Encompass_Preferred
PTC-209 hydrobromide is a small-molecule inhibitor of BMI-1 intended for research use. It has been reported to inhibit BMI-1 activity (IC50 ~0.5 μM in HEK293T cells) and has been used in preclinical studies to impair tumor-initiating cells and affect the tumor microenvironment.
  • Specific BMI-1 inhibitor with reported IC50 ~0.5 μM in HEK293T cells.
  • High purity (99.0%).
  • Solid, light yellow to yellow appearance.
  • Available in small quantities, including a 10 mg vial.
  • Recommended storage: 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-88312 Supplier Medchemexpress LLC Supplier No. HY15888A10MG
PTC-209 hydrobromide is a small-molecule inhibitor of BMI-1 intended for research use. It has been reported to inhibit BMI-1 activity (IC50 ~0.5 μM in HEK293T cells) and has been used in preclinical studies to impair tumor-initiating cells and affect the tumor microenvironment.
  • Specific BMI-1 inhibitor with reported IC50 ~0.5 μM in HEK293T cells.
  • High purity (99.0%).
  • Solid, light yellow to yellow appearance.
  • Available in small quantities, including a 10 mg vial.
  • Recommended storage: 4°C; in solvent: -80°C (6 months) or -20°C (1 month).