Learn More
Medchemexpress LLC HY-124593 5mg , PTC299 CAS:1256565-36-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1245935MG
Encompass_Preferred
HY-124593 5mg PTC299 CAS: 1256565-36-2 PTC299 is an orally active inhibitor of VEGFA mRNA translation that selectively inhibits VEGF protein synthesis at the post-transcriptional level. PTC299 is also a potent inhibitor of dihydroorotate dehydrogenase (DHODH). PTC299 shows good oral bioavailability and lack of off-target kinase inhibition and myelosuppression. PTC299 can be useful for the research of hematologic malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1341