Learn More
Sino Biological Recombinant Human RET Protein (G810R, GST Tag), Active
Catalog No. 502573398
Encompass_Preferred
Recombinant human RET (G810R) (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3398 Supplier Sino Biological Supplier No. R0212VG500
Recombinant human RET (G810R) (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.