Learn More
Medchemexpress LLC PTCDA | 128-69-8 | ≥98.0% | 392.32 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW011009100G
Encompass_Preferred
PTCDA is an organic dye molecule and an organic semiconductor. It is for research use only and not sold to patients.
  • Organic dye molecule
  • Organic semiconductor
Catalog No. 50-004-51709