Learn More
Aviva Systems Biology Corp PTCH1 Antibody - C-terminal region (ARP44249_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44249_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTCH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PTCH1. Alternate names: PTC, BCNS, HPE7, PTC1, PTCH, NBCCS, PTCH11ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53412