Learn More
Anton Paar PTD 80 Peltier Temperature Device for ViscoQC 100 & ViscoQC 300
Catalog No. NC1818588
Encompass_Preferred
A temperature device using Peltier elements and air counter-cooling. Controls sample temperature of DIN / SSA measuring systems in the specified range. Features a fixed CAN cable for power connection and communication with ViscoQC.Use with Measuring System CC12 item NC1818606, CC18 item NC1818604, CC26 item NC1818602, CC12/D18 item NC1818608, SC4-18 item NC1818611, SC4-21 item NC1818613, SC4-27 item NC1818615, DG26 item NC1818619, or DG26/SS NC1818621Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
Catalog No. NC1818588 Supplier Anton Paar Supplier No. 220906
A temperature device using Peltier elements and air counter-cooling. Controls sample temperature of DIN / SSA measuring systems in the specified range. Features a fixed CAN cable for power connection and communication with ViscoQC.Use with Measuring System CC12 item NC1818606, CC18 item NC1818604, CC26 item NC1818602, CC12/D18 item NC1818608, SC4-18 item NC1818611, SC4-21 item NC1818613, SC4-27 item NC1818615, DG26 item NC1818619, or DG26/SS NC1818621Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.