Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PTDA-PEG4-azide | 2639395-40-5 | 99.3% | 539.54 | C22H33N7O9 | 5 MG
SDP

Catalog No. 5000203497
Encompass_Preferred

PTDA-PEG4-azide is a PEG4-linked azide reagent used as an ADC linker and for click-chemistry bioconjugation in research applications. The compound is supplied as a white to off-white oil with high purity and is provided in small, research-scale quantities.

  • High purity (99.3%).
  • Molecular weight 539.54 g/mol.
  • Molecular formula C22H33N7O9.
  • Soluble in DMSO at 125 mg/mL; ultrasonic assistance recommended; use freshly opened DMSO.
  • Storage: pure form -20°C (stable for 3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available in small pack sizes suitable for research use.

Catalog No. 50-002-03497 Supplier Medchemexpress LLC Supplier No. HY1309405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PTDA-PEG4-azide is a PEG4-linked azide reagent used as an ADC linker and for click-chemistry bioconjugation in research applications. The compound is supplied as a white to off-white oil with high purity and is provided in small, research-scale quantities.

  • High purity (99.3%).
  • Molecular weight 539.54 g/mol.
  • Molecular formula C22H33N7O9.
  • Soluble in DMSO at 125 mg/mL; ultrasonic assistance recommended; use freshly opened DMSO.
  • Storage: pure form -20°C (stable for 3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available in small pack sizes suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.