Learn More
Enzo Life Sciences L-alpha-Phosphatidylinositol-3,4,5-P3. 7Na (100µg)
SDP
Supplier:  Enzo Life Sciences BMLPH1070100
Encompass_Preferred
Activates Ca2+-insensitive PKC isotypes δ, ε and η. Binds to GRP1 via a PH domain. Alternative name: PtdIns(3,4,5)P3, (1,2-dipalmitoyl)-, 1,2-Dipalmitoyl-sn-glycero-3-phospho-1-D-myo-inositol-3,4,5-trisphosphate . 7Na. Formulation: Lyophilized. Purity: ≥98% (TLC). Solubility: Soluble in water (10mg/ml) or in chloroform/methanol/water (1:1:0.3) (>10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1434