Learn More
Cayman Chemical PtdIns- 3 4 5-P31-stearoyl 1mg
SDP
Catalog No. 502895982
Encompass_Preferred
A synthetic analog of natural PtdIns featuring C:18 fatty acid in the sn-1 position and arachidonic acid in the sn-2 position of the glycerol backbone; a critical phospholipid species that can served as an anchor for the binding of signal transduction proteins bearing pleckstrin homology (PH) domains
Catalog No. 50-289-5982 Supplier Cayman Chemical Supplier No. 649301 MG
A synthetic analog of natural PtdIns featuring C:18 fatty acid in the sn-1 position and arachidonic acid in the sn-2 position of the glycerol backbone; a critical phospholipid species that can served as an anchor for the binding of signal transduction proteins bearing pleckstrin homology (PH) domains