Learn More
Medchemexpress LLC PtdIns-(345)-P3 (12-dipalmitoyl) (sodium) | 1628353-02-5 | 98.0% | 1138.90 | 100 UG
SDP
Catalog No. 5000360280
Encompass_Preferred
PtdIns-(345)-P3 (12-dipalmitoyl) sodium is a phosphatidylinositol 3,4,5-trisphosphate (PIP3) analog. It can be incorporated into liposomes to establish a backdrop of membrane phospholipids that closely mirrors in vivo conditions. This product is for research use only.
  • Analog of phosphatidylinositol 3,4,5-trisphosphate (PIP3)
  • Can be incorporated into liposomes
  • Establishes a membrane phospholipid environment that closely mimics in vivo conditions
  • For research use only
Catalog No. 50-003-60280 Supplier Medchemexpress LLC Supplier No. HY148971A100UG
PtdIns-(345)-P3 (12-dipalmitoyl) sodium is a phosphatidylinositol 3,4,5-trisphosphate (PIP3) analog. It can be incorporated into liposomes to establish a backdrop of membrane phospholipids that closely mirrors in vivo conditions. This product is for research use only.
  • Analog of phosphatidylinositol 3,4,5-trisphosphate (PIP3)
  • Can be incorporated into liposomes
  • Establishes a membrane phospholipid environment that closely mimics in vivo conditions
  • For research use only