Learn More
Medchemexpress LLC PtdIns-(345)-P3 (12-dipalmitoyl) (sodium) | 1628353-02-5 | ≥98.0% | 1138.90 | 500 UG
SDP
Catalog No. 5000361132
Encompass_Preferred
PtdIns-(345)-P3 (12-dipalmitoyl) sodium (Phosphatidylinositol tris-3,4,5-phosphate, 1,2-dipalmitoyl sodium) is a phosphatidylinositol 3,4,5-trisphosphate (PIP3) analog. It can be incorporated in liposomes to establish a backdrop of membrane phospholipids that closely mirrors in vivo conditions.
  • Phosphatidylinositol 3,4,5-trisphosphate (PIP3) analog
  • Can be incorporated in liposomes
  • Used to establish a backdrop of membrane phospholipids
  • Mimics in vivo conditions
Catalog No. 50-003-61132 Supplier Medchemexpress LLC Supplier No. HY148971A500UG
PtdIns-(345)-P3 (12-dipalmitoyl) sodium (Phosphatidylinositol tris-3,4,5-phosphate, 1,2-dipalmitoyl sodium) is a phosphatidylinositol 3,4,5-trisphosphate (PIP3) analog. It can be incorporated in liposomes to establish a backdrop of membrane phospholipids that closely mirrors in vivo conditions.
  • Phosphatidylinositol 3,4,5-trisphosphate (PIP3) analog
  • Can be incorporated in liposomes
  • Used to establish a backdrop of membrane phospholipids
  • Mimics in vivo conditions