Learn More
Cayman Chemical PtdIns 4 5P2 1 2dipalmitoyl
SDP
Supplier:  Cayman Chemical 100081151 MG
Encompass_Preferred
A synthetic analog of natural PtdIns featuring C16:0 fatty acids at the sn-1 and sn-2 positions; contains the same inositol and DAG stereochemistry as the natural compound
Catalog No. 50-288-9802