Learn More
Cayman Chemical PtdIns 4 5P21 2dIpalmIt 100UG
SDP
Catalog No. 502896256
Encompass_Preferred
A synthetic analog of natural PtdIns featuring C16:0 fatty acids at the sn-1 and sn-2 positions; contains the same inositol and DAG stereochemistry as the natural compound
Catalog No. 50-289-6256 Supplier Cayman Chemical Supplier No. 64924100 UG
A synthetic analog of natural PtdIns featuring C16:0 fatty acids at the sn-1 and sn-2 positions; contains the same inositol and DAG stereochemistry as the natural compound