Learn More
Cayman Chemical PtdIns 5P11 2dIpalmItoy 100UG
SDP
Catalog No. 502896757
Encompass_Preferred
A synthetic analog of natural PtdIns featuring C16:0 fatty acids at the sn-1 and sn-2 positions; contains the same inositol and DAG stereochemistry as that of the natural compound
Catalog No. 50-289-6757 Supplier Cayman Chemical Supplier No. 64925100 UG
A synthetic analog of natural PtdIns featuring C16:0 fatty acids at the sn-1 and sn-2 positions; contains the same inositol and DAG stereochemistry as that of the natural compound