Learn More
Echelon Biosciences Research Labs PTDINS DiC16 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs P0016100UG
Encompass
Phosphatidylinositol diC16 (PI diC16) is a synthetic, purified dipalmitoyl PI.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable
Catalog No. 50-720-553