Learn More
Echelon Biosciences Research Labs PTDINS DiC4 1 mg
SDP
Catalog No. 50720545
Encompass
Phosphatidylinositol diC4 (PI diC4) is a synthetic, purified dibutanoyl PI.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-545 Supplier Echelon Biosciences Research Labs Supplier No. P00041MG
Phosphatidylinositol diC4 (PI diC4) is a synthetic, purified dibutanoyl PI.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable