Learn More
Echelon Biosciences Research Labs PTDINS DiC8 500 ug
SDP
Supplier:  Echelon Biosciences Research Labs P0008500UG
Encompass
Phosphatidylinositol diC8 (PI diC8) is a synthetic, purified dioctanoyl PI.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable
Catalog No. 50-720-550