Learn More
Aviva Systems Biology Corp PTDSS2 Antibody - N-terminal region (ARP49960_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49960_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTDSS2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PTDSS2. Alternate names: FLJ46373, PSS2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-469