Learn More
Diagnostic Biosystems PTEN PTEN (Tumor Suppressor)
Catalog No. 502407505
Encompass_Preferred
PTEN gene is a tumor suppressor gene that maps to chromosome 10q23. PTEN, a novel tumor suppressor, functions as a regulator of both cell cycle progression and apoptosis ). Potentially, mutation and deletion of PTEN gene may result in a new signal transduction pathway related to human malignant tumors. Studies have demonstrated a reduction of PTEN expression in advanced breast cancers
Catalog No. 50-240-7505 Supplier Diagnostic Biosystems Supplier No. PDM574
PTEN gene is a tumor suppressor gene that maps to chromosome 10q23. PTEN, a novel tumor suppressor, functions as a regulator of both cell cycle progression and apoptosis ). Potentially, mutation and deletion of PTEN gene may result in a new signal transduction pathway related to human malignant tumors. Studies have demonstrated a reduction of PTEN expression in advanced breast cancers