Learn More
Enzo Life Sciences PTEN (human), (recombinant) (His-tag) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4020010
Encompass_Preferred
Alternative name: Phosphatase and tensin homolog deleted on chromosome 10. Purity: ≥80% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 150mM sodium chloride, 5mM DTT, 0.03% Brij 35, 0.1mM EDTA and 20% glycerol. Source: Produced in E. coli. Full length human PTEN (aa 1-403) is fused at the N-terminus to a His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2009