Learn More
Biocare Medical, LLC PTEN (Tumor Suppressor) Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM278AK
Encompass_Preferred
PTEN antibody, a novel tumor suppressor, functions as a regulator of both cell cycle progression and apoptosis. Potentially, mutation and deletion of PTEN gene may result in a new signal transduction pathway related to human malignant tumors. Studies have demonstrated a reduction of PTEN expression in advanced breast, prostate and other cancers. In addition, studies also suggest that patients with ErbB2 overexpressing tumors and concurrent low levels of PTEN expression have a poor response to trastuzumab treatment.
Catalog No. 50-826-34