Learn More
ATCC PTEN-CaP8; Prostate Epithelium; Mouse (Mus musculus)
Catalog No. 502382131
Encompass
PTEN-CaP8 is a cell line that was isolated in 2004 from a 10-month-old male mouse. The mouse prostate epithelium (PTEN-P8) cells were infected with a retrovirus carrying Cre and Puromycin-resistant cassette. It can be used for assay development, cancer research, drug development, drug resistance, and high-throughput screening.
Catalog No. 50-238-2131 Supplier ATCC Supplier No. CRL3033
PTEN-CaP8 is a cell line that was isolated in 2004 from a 10-month-old male mouse. The mouse prostate epithelium (PTEN-P8) cells were infected with a retrovirus carrying Cre and Puromycin-resistant cassette. It can be used for assay development, cancer research, drug development, drug resistance, and high-throughput screening.