Learn More
Aviva Systems Biology Corp PTER Antibody - N-terminal region (ARP61159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTER. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PTER. Alternate names: RPR-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-276