Learn More
Cayman Chemical Pterostilbene, Purity: ≥98%, CAS: 537-42-8, 100 mg
SDP
Catalog No. 502955808
Encompass_Preferred
A stilbene with diverse biological activities; reduces acifluorfen-induced photooxidative leakage in C. sativus cotyledon disks at 30 µg/ml; inhibits TPA-induced invasion and migration of HepG2 cells at 25 and 50 µM; inhibits production of NO and PGE2 in LPS-stimulated RAW 264.7 macrophages at 10 and 20 µM; activates PPARα in H4IIEC3 cells at 100 and 300 µM; decreases plasma glucose and LDL levels and increases plasma HDL levels in a hamster model of high-fat diet-induced hypercholesterolemia at 25 ppm in the diet
Catalog No. 50-295-5808 Supplier Cayman Chemical Supplier No. 13000100 MG
A stilbene with diverse biological activities; reduces acifluorfen-induced photooxidative leakage in C. sativus cotyledon disks at 30 µg/ml; inhibits TPA-induced invasion and migration of HepG2 cells at 25 and 50 µM; inhibits production of NO and PGE2 in LPS-stimulated RAW 264.7 macrophages at 10 and 20 µM; activates PPARα in H4IIEC3 cells at 100 and 300 µM; decreases plasma glucose and LDL levels and increases plasma HDL levels in a hamster model of high-fat diet-induced hypercholesterolemia at 25 ppm in the diet