Learn More
Apexbio Technology LLC Pterostilbene(Synonyms: 3,5-Dimethoxy-4'-hydroxy-trans-stilbene, Trans-3,5-dimethoxy-4'-hydroxystilbene), 10mM (in 1mL DMSO), CAS: 537-42-8.
SDP
Catalog No. 5000061310
Encompass_Preferred
Pterostilbene (CAS 537-42-8) is a naturally occurring stilbenoid identified in extracts derived from the plant Pterocarpus indicus Structurally related to resveratrol it demonstrates improved bioavailability and enhanced biological activity Mechanistically pterostilbene exhibits antioxidant anti-inflammatory and antitumor properties primarily mediated through activation of sirtuins and modulation of cell-signaling pathways such as NF- B AMPK and PI3K/Akt Due to these activities pterostilbene is widely utilized in biochemical research exploring cellular aging oxidative stress inflammation and cancer biology Storage is recommended under airtight conditions in cool dry environments
Catalog No. 50-000-61310 Supplier Apexbio Technology LLC Supplier No. N21265.1
Pterostilbene (CAS 537-42-8) is a naturally occurring stilbenoid identified in extracts derived from the plant Pterocarpus indicus Structurally related to resveratrol it demonstrates improved bioavailability and enhanced biological activity Mechanistically pterostilbene exhibits antioxidant anti-inflammatory and antitumor properties primarily mediated through activation of sirtuins and modulation of cell-signaling pathways such as NF- B AMPK and PI3K/Akt Due to these activities pterostilbene is widely utilized in biochemical research exploring cellular aging oxidative stress inflammation and cancer biology Storage is recommended under airtight conditions in cool dry environments